Indazole, 96%
Indazole, 96%
Indazole, 96%
Thermo Scientific Chemicals

Indazole, 96%

CAS: 271-44-3 | C7H6N2 | 118.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122120250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS271-44-3
IUPAC Name1H-indazole
Molecular FormulaC7H6N2
InChI KeyBAXOFTOLAUCFNW-UHFFFAOYSA-N
SMILESN1N=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration with HClO4>=95 %
Melting point146°C to 150°C
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only