Indene, 90%, technical, stabilized
Indene, 90%, technical, stabilized
Indene, 90%, technical, stabilized
Thermo Scientific Chemicals

Indene, 90%, technical, stabilized

CAS: 95-13-6 | C9H8 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 122140010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-13-6
IUPAC Name1H-indene
Molecular FormulaC9H8
InChI KeyYBYIRNPNPLQARY-UHFFFAOYSA-N
SMILESC1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5670 to 1.5750 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
GC>=88 %
Stabilizer5 to 10 ppm tert-Butylcatechol
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only