Indole-3-carbinol, 97%
Indole-3-carbinol, 97%
Indole-3-carbinol, 97%
Thermo Scientific Chemicals

Indole-3-carbinol, 97%

CAS: 700-06-1 | C9H9NO | 147.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12219025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122190250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS700-06-1
IUPAC Name(1H-indol-3-yl)methanol
Molecular FormulaC9H9NO
InChI KeyIVYPNXXAYMYVSP-UHFFFAOYSA-N
SMILESOCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to light orange
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder or flakes
Melting point92°C to 100°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only