Indole-2-carboxylic acid, 99%
Indole-2-carboxylic acid, 99%
Indole-2-carboxylic acid, 99%
Thermo Scientific Chemicals

Indole-2-carboxylic acid, 99%

CAS: 1477-50-5 | C9H7NO2 | 161.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12220010010 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 122200100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1477-50-5
IUPAC Name1H-indole-2-carboxylic acid
Molecular FormulaC9H7NO2
InChI KeyHCUARRIEZVDMPT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=98.5 %
Appearance (Color)Off-white to yellow to brown
Melting point205°C to 209°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only