2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
2-Iodobenzoic acid, 98%
Thermo Scientific Chemicals

2-Iodobenzoic acid, 98%

CAS: 88-67-5 | C7H5IO2 | 248.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 122340250
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.90
Save 4.25
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.90
Save 4.25
Each
Chemical Identifiers
CAS88-67-5
IUPAC Name2-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyCJNZAXGUTKBIHP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to beige
Melting point160°C to 164°C
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only