The PureLink® Air Porous Tape is a component in the PureLink® 96 Plasmid Purification System. The PureLink® 96 Plasmid PurificationRead more
Have Questions?
Catalog Number
Quantity
12262010
also known as 12262-010
50 Pieces
Catalog number 12262010
also known as 12262-010
Price (USD)
109.00
Each
Quantity:
50 Pieces
Price (USD)
109.00
Each
The PureLink® Air Porous Tape is a component in the PureLink® 96 Plasmid Purification System. The PureLink® 96 Plasmid Purification offers a method for high throughput plasmid DNA isolation. The PureLink® Air Porous Tape allows for proper aeration of bacterial cultures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorWhite
For Use With (Application)Automated Nucleic Acid Purification, DNA Extraction