Can I use a low-copy number plasmid for the PureLink 96/384 plasmid purification system?
No. This product is not recommended for use with low-copy number plasmids when the plasmid DNA is for use in automated fluorescent DNA sequencing. Low-copy number plasmids can be used with this system when the samples are for use in PCR amplification. However, keep in mind that the yields from a low-copy plasmid may not be readily visible on a gel.