PureLink™ Pro 96 Viral RNA/DNA Purification Kit - FAQs
PureLink™ Pro 96 Viral RNA/DNA Purification Kit - FAQs
View additional product information for PureLink™ Pro 96 Viral RNA/DNA Purification Kit - FAQs (12280096A)
2 product FAQs found
If I do not use all the wells in the filter plate of the PureLink Pro 96 Purification System, can I use those wells at another time?
Yes. Keep the unused wells covered with Foil Tape. Please note that extra Foil Tape and Receiver Plates will be needed.
My centrifuge does not reach 3,000 x g. For the PureLink Pro 96 system, could I spin at a lower force for a longer time?
Generally speaking, yes. It should work the same if you can keep the same g (rcf) x time.