4-Ketopimelic acid, 98%
4-Ketopimelic acid, 98%
4-Ketopimelic acid, 98%
Thermo Scientific Chemicals

4-Ketopimelic acid, 98%

CAS: 502-50-1 | C7H10O5 | 174.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 122880050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS502-50-1
IUPAC Name4-oxoheptanedioic acid
Molecular FormulaC7H10O5
InChI KeyUDDSEESQRGPVIL-UHFFFAOYSA-N
SMILESOC(=O)CCC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point140°C to 146°C
Titration with NaOH94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only