Sodium thiosulfate pentahydrate, 99+%, extra pure
Sodium thiosulfate pentahydrate, 99+%, extra pure
Sodium thiosulfate pentahydrate, 99+%, extra pure
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99+%, extra pure

CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 123370025
Price (EUR)
78,40
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
78,40
Each
Chemical Identifiers
CAS10102-17-7
IUPAC Namedisodium pentahydrate sulfanidesulfonate
Molecular FormulaH10Na2O8S2
InChI KeyPODWXQQNRWNDGD-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
pH6 to 8.4 (10 % at 20°C)
Calcium (Ca)=<50 ppm
Chloride (Cl)=<500 ppm
Sulfur (S)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only