2-Pentanone, 99%, pure
2-Pentanone, 99%, pure
2-Pentanone, 99%, pure
Thermo Scientific Chemicals

2-Pentanone, 99%, pure

CAS: 107-87-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 123500010
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
114.00
Save 17.35
Each
Chemical Identifiers
CAS107-87-9
IUPAC Namepentan-2-one
Molecular FormulaC5H10O
InChI KeyXNLICIUVMPYHGG-UHFFFAOYSA-N
SMILESCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3890 to 1.3910 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only