Platinum™ SuperFi™ DNA Polymerase, 5 x 500 units - FAQs
Platinum™ SuperFi™ DNA Polymerase, 5 x 500 units - FAQs
View additional product information for Platinum™ SuperFi™ DNA Polymerase - FAQs (12351250, 12351050, 12351010)
1 product FAQs found
Can I use Taq polymerase to generate my gene of interest for directional TOPO cloning?
No, your gene of interest must be amplified with a proofreading polymerase such as Platinum SuperFi DNA Polymerase or AccuPrime Pfx DNA Polymerase that leaves blunt ends for directional TOPO cloning.