Sodium thiosulfate, 0.1 N standard solution
Sodium thiosulfate, 0.1 N standard solution
Sodium thiosulfate, 0.1 N standard solution
Acros Organics

Sodium thiosulfate, 0.1 N standard solution

CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
View all
Plastic bottle
Catalog number 124270010
also known as 12427
HSN 28323090
Price (INR)
6,542.00
Each
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
6,542.00
Each
Chemical Identifiers
CAS7772-98-7
IUPAC Namedisodium sulfanidesulfonate
Molecular FormulaNa2O3S2
InChI KeyAKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Titration Iodimetric0.0950 to 0.1050 N (20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent, Volumetric standard solution
RUO – Research Use Only