Oxalic acid, 0.1 N standard solution
Oxalic acid, 0.1 N standard solution
Oxalic acid, 0.1 N standard solution
Thermo Scientific Chemicals

Oxalic acid, 0.1 N standard solution

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number 124280010
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.20
Save 8.55
Each
Chemical Identifiers
CAS144-62-7
IUPAC Nameoxalic acid
Molecular FormulaC2H2O4
InChI KeyMUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.0998 to 0.1002 N (20°C)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only