IMDM
Gibco™

IMDM

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
12440053500 mL
1244006110 x 500 mL
124400461000 mL
Catalog number 12440053
Price (EUR)
24,65
Online exclusive
29,04
Save 4,39
Each
Quantity:
500 mL
Customize this product
Price (EUR)
24,65
Online exclusive
29,04
Save 4,39
Each

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: L-glutamine, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineJurkat, COS-7, and macrophage cells
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeIMDM (Iscove's Modified Dulbecco's Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesHigh Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Without AdditivesNo α-Thioglycerol, No 2-Mercaptoethanol
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

What is the shelf life of the DMEM, high glucose, pyruvate medium once the bottle is opened and the medium is supplemented?

We do not provide stability data for the product once it is opened as it would depend on the usage and storage conditions.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to store DMEM, high glucose, pyruvate medium in the dark?

Yes, the medium should be stored in the dark because there are some components in the medium such as HEPES, Tryptophan, and Riboflavin, etc. that are sensitive to light.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Ex vivo differentiation of cord blood stem cells into megakaryocytes and platelets.
Authors:Pineault N, Robert A, Cortin V, Boyer L
Journal:Methods Mol Biol
PubMed ID:23179834
Megakaryocytes (MK) are hematopoietic cells present in the bone marrow that are responsible for the production and release of platelets in the circulation. Given their very low frequency (<1%), human MK often need to be derived in culture to study their development or to generate sufficient material for biological studies. ... More