Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals

数量:
500 mL
1 L
2.5 L
25 L
パッケージ:
すべて表示
Plastic bottle
Glass Bottle
製品番号(カタログ番号) 124665001
価格(JPY)
-
数量:
500 mL
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
さらに表示
Chloride (Cl)=<0.5 ppm
Copper (Cu)=<0.05 ppm
Cadmium (Cd)=<0.01 ppm
Arsenic (As)=<0.01 ppm
Appearance (Form)Liquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only