Calcium carbonate, 98+%, pure, heavy powder
Calcium carbonate, 98+%, pure, heavy powder
Calcium carbonate, 98+%, pure, heavy powder
Thermo Scientific Chemicals

Calcium carbonate, 98+%, pure, heavy powder

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 124670010
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.90
Save 13.25
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 28-Dec-2026
87.90
Save 13.25
Each
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric98.0 to 102.0 %
Appearance (Color)White to almost white to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only