Advanced MEM
Gibco™

Advanced MEM

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reducedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1249202110 x 500 mL
12492013500 mL
Catalog number 12492021
Price (USD)
440.65
Online exclusive
512.00
Save 71.35
Each
Quantity:
10 x 500 mL
Customize this product
Price (USD)
440.65
Online exclusive
512.00
Save 71.35
Each

Advanced MEM (Minimum Essential medium) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic MEM, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced DMEM / F-12, with no adaptation, include PK-15, MDCK, Vero, WI-38 and Hep-2.

This Advanced MEM is manufactured as follows:

With: Glucose, Non Essential Amino Acids, Sodium pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced MEM is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced MEM requires supplementation with 1-5% Fetal Bovine Serum and 4mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iPaul Price, Brad Stiles, David Staines, Ethel Evege, Femi Fatunmbi, Kate Wagner, Lori Nestler, David Jayme. Advanced Media as Alternatives to Classical Basal Media. FOCUS, 2003. 25.2:3-6.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineHeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMEM (Minimum Essential Medium)
Quantity10 x 500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionAmbient
ClassificationAnimal Origin
FormLiquid
Serum LevelReduced Serum
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesLow Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
Without AdditivesNo Glutamine, No HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Where can I find the osmolality for MEM Medium?

The osmolality is listed in the COA for the particular lot number of the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.