1-(4-Nitrophenyl)piperazine, 98%
1-(4-Nitrophenyl)piperazine, 98%
1-(4-Nitrophenyl)piperazine, 98%
Thermo Scientific Chemicals

1-(4-Nitrophenyl)piperazine, 98%

CAS: 6269-89-2 | C10H13N3O2 | 207.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 124941000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6269-89-2
IUPAC Name1-(4-nitrophenyl)piperazine
Molecular FormulaC10H13N3O2
InChI KeyVWOJSRICSKDKAW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point130°C to 134°C
Appearance (Color)Yellow to ochre
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only