DL-Lactic acid, 90%
DL-Lactic acid, 90%
DL-Lactic acid, 90%
Thermo Scientific Chemicals

DL-Lactic acid, 90%

CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 125060010
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 28-Dec-2026
159.00
Save 23.35
Each
Chemical Identifiers
CAS50-21-5
IUPAC Name2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILESCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4252 to 1.4272 (20°C, 589 nm)
Acids(Citric,oxalic,phosphoric or tartaric) Passes test
Appearance (Form)Liquid
Residue after ignition=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only