Malonyl dichloride, 97%
Malonyl dichloride, 97%
Malonyl dichloride, 97%
Thermo Scientific Chemicals

Malonyl dichloride, 97%

CAS: 1663-67-8 | C3H2Cl2O2 | 140.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12528025025 gGlass bottle
Catalog number 125280250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1663-67-8
IUPAC Namepropanedioyl dichloride
Molecular FormulaC3H2Cl2O2
InChI KeySXYFKXOFMCIXQW-UHFFFAOYSA-N
SMILESClC(=O)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange to brown
Infrared spectrumConforms
Refractive index1.4620 to 1.4660 (20°C, 589 nm)
Appearance (Form)Liquid
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only