CD 293 AGT™ Medium, 10 L - FAQs
CD 293 AGT™ Medium, 10 L - FAQs
View additional product information for CD 293 AGT™ Medium - FAQs (12529003, 12529001, 12529012, 12529020)
3 product FAQs found
Can you please provide information as to how many grams of powder are present in your CD 293 AGT Medium products?
1 L (Cat. No. 12529020): 17.9 g/L
10 L (Cat. No. 12529012): 178.8 g/10 L
100 L (Cat. No. 12529001): 1788.4 g/100 L
10 kg (Cat. No. 12529003): 10,000 g/10 kg
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What are the main benefits of using AGT?
Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations
Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is AGT?
Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.
AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.