L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
Thermo Scientific Chemicals

L(-)-Menthol, 99.5%

CAS: 2216-51-5 | C10H20O | 156.27 g/mol
数量:
5 g
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 125400025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2216-51-5
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyNOOLISFMXDJSKH-KXUCPTDWSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
さらに表示
Infrared spectrumConforms
GC>=99.4 %
Specific optical rotation-48° to -51° (20°C, 589 nm) (c=10, EtOH)
Appearance (Form)Crystals or crystalline needles
Appearance (Color)Colorless to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only