2-Mercaptobenzothiazole, 95%
2-Mercaptobenzothiazole, 95%
2-Mercaptobenzothiazole, 95%
Thermo Scientific Chemicals

2-Mercaptobenzothiazole, 95%

CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 125450010
Price (ZAR)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS149-30-4
IUPAC Name2,3-dihydro-1,3-benzothiazole-2-thione
Molecular FormulaC7H5NS2
InChI KeyYXIWHUQXZSMYRE-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point172°C to 180°C
Titration with NaOH>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only