2-Mercaptoethyl sulfide, 90%
2-Mercaptoethyl sulfide, 90%
2-Mercaptoethyl sulfide, 90%
Thermo Scientific Chemicals

2-Mercaptoethyl sulfide, 90%

CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 125490100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3570-55-6
IUPAC Name2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
Molecular FormulaC4H10S3
InChI KeyKSJBMDCFYZKAFH-UHFFFAOYSA-N
SMILESSCCSCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5900 to 1.5980 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only