Can you offer an alternative product for Superfrost Plus Microscope Slides (Cat. No. 1255015)?
The Fisherbrand Superfrost Plus Microscope Slides are available from Fisher Scientific, linked below:
Fisherbrand Superfrost Plus Microscope Slides (Cat. Nos. 22-037-246, 12-550-15)
Fisherbrand Superfrost Plus Microscope Slides (Cat. Nos. 22-037-246, 12-550-15)