Mesityl oxide, 99%, mixture of alpha- and beta-isomers
Mesityl oxide, 99%, mixture of alpha- and beta-isomers
Mesityl oxide, 99%, mixture of alpha- and beta-isomers
Thermo Scientific Chemicals

Mesityl oxide, 99%, mixture of alpha- and beta-isomers

CAS: 141-79-7 | C6H10O | 98.14 g/mol
数量:
1 L
2.5 L
10 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 125600010
価格(JPY)
-
数量:
1 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS141-79-7
IUPAC Name4-methylpent-3-en-2-one
Molecular FormulaC6H10O
InChI KeySHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILESCC(C)=CC(C)=O
さらに表示
Appearance (Color)Clear colorless to yellow
Refractive index1.4420 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only