Gibco™ TrypLE™ Select Enzyme (1X), no phenol red, 10 L - FAQs
Gibco™ TrypLE™ Select Enzyme (1X), no phenol red, 10 L - FAQs
View additional product information for TrypLE™ Select Enzyme (1X), no phenol red - FAQs (12563029, 12563011, 12563030, 12563031)
1 product FAQs found
Can I use TrypLE Select Enzyme to detach cells from glass surfaces?
Yes, TrypLE Select Enzyme can detach cells from glass surfaces.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.