What is the molecular weight of insulin in Insulin, human recombinant, zinc solution (Cat. No. 12585014)?
The molecular weight of insulin in Insulin, human recombinant, zinc solution (Cat. No. 12585014) is 5807.7 g/mol.