4-Methylbenzophenone, 97%
4-Methylbenzophenone, 97%
4-Methylbenzophenone, 97%
Thermo Scientific Chemicals

4-Methylbenzophenone, 97%

CAS: 134-84-9 | C14H12O | 196.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 126352500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS134-84-9
IUPAC Name(4-methylphenyl)(phenyl)methanone
Molecular FormulaC14H12O
InChI KeyWXPWZZHELZEVPO-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only