4-Methylbenzyl cyanide, 98%
4-Methylbenzyl cyanide, 98%
4-Methylbenzyl cyanide, 98%
Thermo Scientific Chemicals

4-Methylbenzyl cyanide, 98%

CAS: 2947-61-7 | C9H9N | 131.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 126440050
Price (USD)
29.10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.10
Each
Chemical Identifiers
CAS2947-61-7
IUPAC Name2-(4-methylphenyl)acetonitrile
Molecular FormulaC9H9N
InChI KeyRNHKXHKUKJXLAU-UHFFFAOYSA-N
SMILESCC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5170 to 1.5195 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only