Methyl chloroformate, 99%
Methyl chloroformate, 99%
Methyl chloroformate, 99%
Thermo Scientific Chemicals

Methyl chloroformate, 99%

CAS: 79-22-1 | C2H3ClO2 | 94.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 126531000
Price (USD)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79-22-1
IUPAC Namemethyl carbonochloridate
Molecular FormulaC2H3ClO2
InChI KeyXMJHPCRAQCTCFT-UHFFFAOYSA-N
SMILESCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only