Sf-900™ III SFM, 500 mL - FAQs
Sf-900™ III SFM, 500 mL - FAQs
View additional product information for Sf-900™ III SFM - FAQs (12658035, 12658001, 12658027, 12658019)
3 product FAQs found
What antimicrobials can be used in insect culture and at what concentration?
Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is it necessary to heat-inactivate my serum before adding it to my medium?
Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.
Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer serum-free insect media?
Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.