1-Methylcyclohexanol, 96%
1-Methylcyclohexanol, 96%
1-Methylcyclohexanol, 96%
Thermo Scientific Chemicals

1-Methylcyclohexanol, 96%

CAS: 590-67-0 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 126601000
Price (USD)
288.65
Special offer
Online exclusive
Ends: 28-Dec-2026
339.00
Save 50.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
288.65
Special offer
Online exclusive
Ends: 28-Dec-2026
339.00
Save 50.35
Each
Chemical Identifiers
CAS590-67-0
IUPAC Name1-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyVTBOTOBFGSVRMA-UHFFFAOYSA-N
SMILESCC1(O)CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Form)Liquid after melting
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only