N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 126720025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS105-59-9
IUPAC Name2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Molecular FormulaC5H13NO2
InChI KeyCRVGTESFCCXCTH-UHFFFAOYSA-N
SMILESCN(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4675 to 1.4695 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.5 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only