Methyl methacrylate, 99%, stabilized
Methyl methacrylate, 99%, stabilized
Methyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals

Methyl methacrylate, 99%, stabilized

Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L | CAS: 80-62-6 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 127140010
Price (USD)
84.80
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
84.80
Each
Chemical Identifiers
CAS80-62-6
IUPAC Namemethyl 2-methylprop-2-enoate
Molecular FormulaC5H8O2
InChI KeyVVQNEPGJFQJSBK-UHFFFAOYSA-N
SMILESCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Acidity=<0.005 % (as methacrylic acid)
GC>=98.5 %
Refractive index1.4130 to 1.4160 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only