4-Methyl-3-nitroaniline, 97%
4-Methyl-3-nitroaniline, 97%
4-Methyl-3-nitroaniline, 97%
Thermo Scientific Chemicals

4-Methyl-3-nitroaniline, 97%

CAS: 119-32-4 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 127250050
Price (USD)
27.70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
27.70
Each
Chemical Identifiers
CAS119-32-4
IUPAC Name4-methyl-3-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyGDIIPKWHAQGCJF-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Melting point74°C to 78°C
Appearance (Color)Ochre to orange to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only