MesenPRO RS™ Medium - FAQs
MesenPRO RS™ Medium - FAQs
View additional product information for MesenPRO RS™ Medium - FAQs (12746012)
5 product FAQs found
Can I use MesenPRO RS Medium with both StemPro Human Adipose-Derived Stem Cells and StemPro Bone Marrow (BM) Mesenchymal Stem Cells?
Yes, the MesenPRO RS Medium (with growth supplement added) can be used for both cell types.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I culture mesenchymal stem cells (MSCs) in vitro?
MSCs may be cultured in either serum-containing medium or serum-free medium. The standard culturing conditions are DMEM, low glucose with 10% FBS. MSCs can also be grown in reduced-serum (2%) Gibco MesenPRO RS Medium (Cat. No. 12746-012), or in serum-free Gibco StemPro MSC SFM XenoFree Medium (Cat. No. A10675) and Gibco StemPro MSC SFM (Cat. No. A10332). In general, MSCs grow better under hypoxic conditions (2% O2).
How do you characterize human mesenchymal stem cells (MSCs)?
Human MSCs can be identified by flow cytometry, typically displaying CD73+, CD90+, CD105+, CD11b-, and CD45- marker characteristics (Blood 109:4245 (2007)). Human MSC expresses Oct4, Sox2, and Rex-1; these may be verified using RT-PCR (Arthritis Res Ther 9:204 (2007)).
What are mesenchymal stem cells?
Mesenchymal stem cells (MSCs) are multipotent cells isolated primarily from bone marrow or fat tissues that exhibit the ability to differentiate into bone, cartilage, and fat cells. Under normal cell culture conditions, MSCs isolated from bone marrow are spindle shaped with the unique ability to adhere to uncoated plastic culture dishes (Arthritis Res Ther 9:204 (2007)).