Product Image
Gibco™

HuMEC Supplement Kit

HuMEC Supplement Kit
Have Questions?
Catalog NumberQuantity
127550131 Kit
Catalog number 12755013
Price (USD)
156.00
Each
Quantity:
1 Kit
Price (USD)
156.00
Each

HuMEC Supplement Kit contains growth factors and other components and is designed to work with HuMEC Basal Serum-Free Medium (12753018). Together, these products are engineered specifically for the culture of human mammary epithelial cells (A10565). This translates to greater experimental validity, more reproducible results, and reduction of lot-to-lot variability and aberrations that can occur with the addition of serum. Human mammary epithelial cells cultured in HuMEC Medium have gene expression profiles consistent with normal mammary epithelial cells, as tested on a cDNA microarray.

Applications:
HuMEC Medium is optimized specifically for the growth of human mammary epithelial cells, and has been performance tested in a growth assay using normal human mammary epithelial cells. HuMEC Medium promotes superior growth of normal human mammary epithelial cells, making it ideal for researchers in breast cancer, drug discovery, and other aspects of breast biology.

Contents and Storage:
The HuMEC Supplement Kit includes 5 ml of a supplement mix containing epidermal growth factor, hydrocortisone, isoproterenol, transferrin, and insulin, and 25 mg of bovine pituitary extract. Store the supplement mix at -5°C to -20°C.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cell TypeEpithelial Cells (Mammary)
Culture TypeMammalian Cell Culture
Quantity1 Kit
Serum LevelSerum-Free
Shelf Life18 Months
SpeciesHuman
Product TypeHuMEC Supplement
SterilitySterile-filtered
Unit SizeEach