Product Image
Thermo Scientific™

β-Glucuronidase Enzyme

Use this enzyme with the Thermo Scientific™ CEDIA™ Benzodizepine Assay for enhanced sensitivity of parent drug.
Have Questions?
Catalog NumberQuantity
1276801 x 5 mL
Catalog number 127680
Price (USD)
-
Quantity:
1 x 5 mL

Use the ß-Glucuronidase Enzyme with the Thermo Scientific™ CEDIA™ Benzodizepine Assay for enhanced sensitivity of parent drug by hydrolyzing the glucuronide metabolites. Hydrolysis removes the glucuronide part of the molecule so the parent drug can be detected by the antibody in the assay.

    • Increased sensitivity for the detection of Benzodiazepines
    • For hydrolysis of the glucuronide metabolites
For In Vitro Diagnostic Use with the CEDIA™ Benzodiazepine Assay Only
Available in select countries only.
Specifications
Descriptionβ-Glucuronidase Enzyme - For In Vitro Diagnostic use with the CEDIA™ Benzodiazepine Assay Only
Formulationß-glucuronidase, extracted from E. coli K 12, in 50% glycerol
Quantity1 x 5 mL
Sample TypeUrine
Stability2° to 8°C until the expiration date printed on the label
TypeHydrolysis Reagent
pHApproximately 6.5
Unit SizeEach
Contents & Storage
-15°C to -25°C