mono-Methyl succinate, 95%
mono-Methyl succinate, 95%
mono-Methyl succinate, 95%
Thermo Scientific Chemicals

mono-Methyl succinate, 95%

CAS: 3878-55-5 | C5H8O4 | 132.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 127730100
Price (USD)
59.60
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.60
Each
Chemical Identifiers
CAS3878-55-5
IUPAC Name4-methoxy-4-oxobutanoic acid
Molecular FormulaC5H8O4
InChI KeyJDRMYOQETPMYQX-UHFFFAOYSA-N
SMILESCOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Form)Crystals and/or chunks
Infrared spectrumConforms
Melting point52°C to 58°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only