Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.7%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
1277900505LPlastic drum
127791000100mLGlass bottle
1277900101LGlass bottle
1277900252.5LGlass bottle
12779025025LPlastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127790050
Price (EUR)
128,00
Each
Quantity:
5L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
128,00
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<20 APHA
GC>=99.6 %
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only