Luria Broth Base (Miller's LB Broth Base), powder
Invitrogen™

Luria Broth Base (Miller's LB Broth Base), powder

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The originalRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
127950842.5 kg
12795027500 g
Catalog number 12795084
Price (USD)
451.65
Online exclusive
480.00
Save 28.35
Each
Quantity:
2.5 kg
Price (USD)
451.65
Online exclusive
480.00
Save 28.35
Each

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original formulation published in 1951 by Guiseppe Bertani to study phage plaque formation and named Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Widely used LB medium formulations differ in salt concentration:
Miller: 10 g/L NaCl
Lennox: 5 g/L NaCl
Luria: 0.5 g/L NaCl

Luria Broth Base (Miller's formulation) is recommended for the general maintenance and growth of E. coli as it allows for fast cell expansion and supports high cell density. In cases where salt-sensitive antibiotics are being used for selection (such as Zeocin, tetracycline or chloramphenicol), Lennox formulation medium would be recommended.

Luria Broth Base formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, and 10 g NaCl.
To prepare 1 L of Miller-LB media, add 25 g of powder to 1 L distilled water and mix by vortexing. Adjust the pH to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C.

Find the media format that fits your needs
We offer other ready-to-use media formulations, such as LB Broth, Terrific Broth or S.O.C recovery medium.
Explore other media formats for bacterial growth.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatBottle
Media TypeLB Broth Base (Miller's LB Broth Base)
Preparation MethodAutoclave
Quantity2.5 kg
Shipping ConditionRoom Temperature
Product TypeDehydrated Culture Media
SpeciesEscherichia coli
Target Organism ClassColiforms
Unit SizeEach
Contents & Storage
• 2.5 kg LB Broth Dehydrated Media Powder

Store at room temperature.

Frequently asked questions (FAQs)

Do you offer an alternative for Luria Broth Base (Miller's LB Broth Base), powder (Cat. No. 12795027, 12795084)?

We do not offer any alternatives for this product.