5-Methyl-DL-tryptophan, 98%
5-Methyl-DL-tryptophan, 98%
5-Methyl-DL-tryptophan, 98%
Thermo Scientific Chemicals

5-Methyl-DL-tryptophan, 98%

CAS: 951-55-3 | C12H14N2O2 | 218.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 127972500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS951-55-3
IUPAC Name2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
Molecular FormulaC12H14N2O2
InChI KeyHUNCSWANZMJLPM-UHFFFAOYSA-N
SMILESCC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals (as Pb)=<10 ppm
Appearance of solution(5% soln. in 1 N NaOH) clear, pale yellowish
Infrared spectrumConforms
Water=<1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only