Myrcene, 90%, tech., stabilized
Myrcene, 90%, tech., stabilized
Myrcene, 90%, tech., stabilized
Thermo Scientific Chemicals

Myrcene, 90%, tech., stabilized

CAS: 123-35-3 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 128080025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS123-35-3
IUPAC Name7-methyl-3-methylideneocta-1,6-diene
Molecular FormulaC10H16
InChI KeyUAHWPYUMFXYFJY-UHFFFAOYSA-N
SMILESCC(C)=CCCC(=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=88 %
Appearance (Form)Liquid
Refractive index1.4660 to 1.4720 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only