1-Naphthoic acid, 98%
1-Naphthoic acid, 98%
1-Naphthoic acid, 98%
Thermo Scientific Chemicals

1-Naphthoic acid, 98%

CAS: 86-55-5 | C11H8O2 | 172.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
128181000100 gPlastic bottle
Catalog number 128181000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS86-55-5
IUPAC Namenaphthalene-1-carboxylic acid
Molecular FormulaC11H8O2
InChI KeyLNETULKMXZVUST-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)Light yellow to brown
Melting point160°C to 164°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only