m-Nitroacetophenone, 98%
m-Nitroacetophenone, 98%
m-Nitroacetophenone, 98%
Thermo Scientific Chemicals

m-Nitroacetophenone, 98%

CAS: 121-89-1 | C8H7NO3 | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 128331000
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.80
Save 5.15
Each
Chemical Identifiers
CAS121-89-1
IUPAC Name1-(3-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeyARKIFHPFTHVKDT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point76°C to 80°C
Appearance (Color)Yellow to yellow-green to yellow-brown
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only