2-Nitroaniline, 98%
2-Nitroaniline, 98%
2-Nitroaniline, 98%
Thermo Scientific Chemicals

2-Nitroaniline, 98%

CAS: 88-74-4 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number 128355000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-74-4
IUPAC Name2-nitroaniline
Molecular FormulaC6H6N2O2
InChI KeyDPJCXCZTLWNFOH-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystals or flakes
GC>=97.5 %
Appearance (Color)Orange to brown
Melting point70°C to 74°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only