3-Nitrobenzoic acid, 99%
3-Nitrobenzoic acid, 99%
3-Nitrobenzoic acid, 99%
Thermo Scientific Chemicals

3-Nitrobenzoic acid, 99%

CAS: 121-92-6 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Catalog number 128455000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS121-92-6
IUPAC Name3-nitrobenzoic acid
Molecular FormulaC7H5NO4
InChI KeyAFPHTEQTJZKQAQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)White to light yellow or light green
Melting point139°C to 143°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only