2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals
2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals
2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
12847010010 gGlass bottle
Catalog number 128470100
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Chemical Identifiers
CAS612-24-8
IUPAC Name2-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeySWBDKCMOLSUXRH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point107°C to 112°C
GC>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only